Sodium 1-tetradecanesulfonate
Supplier: TCI America
Synonyms:
Sodium tetradecane-1-sulphonate, Sodium tetradecane-1-sulfonate
T0757-1G
T0757-10G
TCT0757-001GEA
91.64
CAD
TCT0757-001G
TCT0757-010G
Sodium 1-tetradecanesulfonate
Sodium 1-tetradecanesulphonate
CAS Number: 6994-45-2
MDL Number: MFCD00043415
Molecular Formula: C14H30O3S
Molecular Weight: 300.43
Form: Crystal
Color: White
Melting point (°C): 227
Other Product Names / Synonyms: 1-Tetradecanesulfonic Acid Sodium Salt, Tetradecylsulfonic Acid Sodium Salt
Formula:
C₁₄H₂₉NaO₃S MW: 300.430 g/mol Melting Pt: 227 °C |
MDL Number:
MFCD00043415 CAS Number: 6994-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...