2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture) ≥95.0%

Supplier: TCI America

T0724-25G T0724-500G
TCT0724-025GEA 41.08 CAD
TCT0724-025G TCT0724-500G
2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture) ≥95.0%
2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso-mixture)

CAS Number: 126-86-3
MDL Number: MFCD00008942
Molecular Formula: C14H26O2
Molecular Weight: 226.36
Purity/Analysis Method: >95.0% (GC)
Form: Wax
Color: White
Boiling point (°C): 260
Melting point (°C): 54
Flash Point (°C): 113


Other Product Names / Synonyms: 4,7-Dihydroxy-2,4,7,9-tetramethyl-5-decyne

Formula: C₁₄H₂₆O₂
Boiling Pt: 260 °C
Melting Pt: 54 °C
Flash Pt: 113 °C
MDL Number: MFCD00008942
CAS Number: 126-86-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0c023fdaa688749b60b93f2922f8257ce8d443c&vid=a0c023fdaa688749b60b93f2922f8257ce8d443c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR