L(+)-Diisopropyl tartrate ≥98.0%

Supplier: TCI America

T0621-500G T0621-25G
TCT0621-500GEA 432.92 CAD
TCT0621-500G TCT0621-025G
L(+)-Diisopropyl tartrate ≥98.0%
L(+)-Diisopropyl tartrate

CAS Number: 2217-15-4
MDL Number: MFCD00064450
Molecular Formula: C10H18O6
Molecular Weight: 234.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 275
Flash Point (°C): 110
Specific Gravity (20/20): 1.12
Specific rotation [a]20/D: 17 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-(+)-Tartaric Acid Diisopropyl Ester

Formula: C₁₀H₁₈O₆
Boiling Pt: 275 °C
Density: 1.12 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00064450
CAS Number: 2217-15-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a6d7441f49d81a0884f8b39c2f25a48bc8397c7&vid=7a6d7441f49d81a0884f8b39c2f25a48bc8397c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR