D-(+)-Turanose ≥98.0% (by HPLC)

Supplier: TCI America

T0542-1G T0542-5G
TCT0542-001GEA 105.86 CAD
TCT0542-001G TCT0542-5G
D-(+)-Turanose ≥98.0% (by HPLC)
D(+)-Turanose

CAS Number: 547-25-1
MDL Number: MFCD00006606
Molecular Formula: C12H22O11
Molecular Weight: 342.30
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Specific rotation [a]20/D: 75.5 deg (C=4, H2O)

Formula: C₁₂H₂₂O₁₁
MDL Number: MFCD00006606
CAS Number: 547-25-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2d2ad3a2bd64c401d372009ca8891c9bf046c1d&vid=f2d2ad3a2bd64c401d372009ca8891c9bf046c1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR