Tripropylene glycol (mixture of isomers) ≥98.0%

Supplier: TCI America

Synonyms: Tripropylene glycol

T0523-25ML T0523-500ML
TCT0523-025MLEA 28.44 CAD
TCT0523-025ML TCT0523-500ML
Tripropylene glycol (mixture of isomers) ≥98.0%
Tripropylene glycol (mixture of isomers)

CAS Number: 24800-44-0
MDL Number: MFCD00014405
Molecular Formula: C9H20O4
Molecular Weight: 192.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 146
Specific Gravity (20/20): 1.02

Formula: C₉H₂₀O₄
MW: 192.26 g/mol
Density: 1.02 (20 °C)
Flash Pt: 146 °C
MDL Number: MFCD00014405
CAS Number: 24800-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fa78b57cd02e6bb9aec6c541f777a78f1532aec&vid=8fa78b57cd02e6bb9aec6c541f777a78f1532aec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR