2,4,6-Trimethylstyrene ≥97.0% stabilized
Supplier: TCI America
T0493-5ML
T0493-1ML
TCT0493-005MLEA
268.6
CAD
TCT0493-005ML
TCT0493-001ML
2,4,6-Trimethylstyrene ≥97.0% stabilized
2,4,6-Trimethylstyrene
(stabilized with TBC)
CAS Number: 769-25-5
MDL Number: MFCD00008613
Molecular Formula: C11H14
Molecular Weight: 146.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 206
Flash Point (°C): 75
Specific Gravity (20/20): 0.91
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3,5-Trimethyl-2-vinylbenzene, 2-Vinylmesitylene
Formula:
C₁₁H₁₄ Boiling Pt: 206 °C Density: 0.91 (20 °C) Flash Pt: 75 °C |
MDL Number:
MFCD00008613 CAS Number: 769-25-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...