Trimethylamine-N-oxide dihydrate ≥98.0%

Supplier: TCI America

Synonyms: Trimethylamine oxide dihydrate

T0466-25G T0466-500G T0466-100G
TCT0466-025GEA 114.53 CAD
TCT0466-025G TCT0466-500G TCT0466-100G
Trimethylamine-N-oxide dihydrate ≥98.0%
Trimethylamine N-oxide dihydrate

CAS Number: 62637-93-8
MDL Number: MFCD00002048
Molecular Formula: C3H9NO
Molecular Weight: 75.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 95

Formula: C₃H₉NO·2H₂O
MW: 111.14 g/mol (75.11 anhy)
Melting Pt: 95 °C
MDL Number: MFCD00149077
CAS Number: 62637-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=709b62d620052f88591e91ac90efe6bc1c46ce28&vid=709b62d620052f88591e91ac90efe6bc1c46ce28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR