Phenylfluorone
Supplier: TCI America
Synonyms:
9-Phenyl-2,3,7-trihydroxy-6-fluorone, 9-Phenyl-3-fluorone, 2,6,7-Trihydroxy-9-phenyl-3-isoxanthone
T0450-1G
T0450-5G
TCT0450-001GEA
34.76
CAD
TCT0450-001G
TCT0450-5G
Phenylfluorone
Phenylfluorone
CAS Number: 975-17-7
MDL Number: MFCD00005048
Molecular Formula: C19H12O5
Molecular Weight: 320.30
Form: Crystal
Color: Pale Yellow
Other Product Names / Synonyms: 9-Phenyl-2,3,7-trihydroxy-6-fluorone, 2,6,7-Trihydroxy-9-phenyl-3-isoxanthone
Formula:
C₁₉H₁₂O₅ MW: 320.30 g/mol |
MDL Number:
MFCD00005048 CAS Number: 975-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...