p-Toluenesulfinic Acid Sodium Salt ≥98.0%
Supplier: TCI America
Synonyms:
Sodium 4-methylbenzenesulphinate, Sodium-p-toluenesulphinate, Sodium 4-methylbenzenesulfinate, Sodiumtoluene-4-sulfinate, Sodium-p-toluenesulfinate, 4-Methylbenzenesulphinic acid sodium salt, 4-Methylbenzenesulfinic acid sodium salt, Sodiumtoluene-4-sulphinate
CAS Number: 824-79-3
MDL Number: MFCD00013136
Molecular Formula: C7H8O2S
Molecular Weight: 178.18
Purity/Analysis Method:
Form: Crystal
Color: White
Formula:
C₇H₇NaO₂S Melting Pt: 300 °C |
MDL Number:
MFCD00013136 CAS Number: 824-79-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...