Tetrazolium violet ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T0174-1G T0174-10G
TCT0174-001GEA 65.87 CAD
TCT0174-001G TCT0174-010G
Tetrazolium violet ≥98.0% (by titrimetric analysis)
Tetrazolium Violet

CAS Number: 1719-71-7
MDL Number: MFCD00011875
Molecular Formula: C23H17ClN4
Molecular Weight: 384.87
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Very Pale Yellow


Other Product Names / Synonyms: 2,5-Diphenyl-3-(1-naphthyl)tetrazolium Chloride, Violet Tetrazolium

Formula: C₂₃H₁₇ClN₄
MDL Number: MFCD00011875
CAS Number: 1719-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=510e1faf3bd65a1c653ad1243e7354500c93c7a8&vid=510e1faf3bd65a1c653ad1243e7354500c93c7a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR