Fisetin ≥96.0%

Supplier: TCI America

Synonyms: 3,7,3',4'-Tetrahydroxyflavone

T0121-100MG T0121-1G T0121-5G
TCT0121-100MGEA 79.57 CAD
TCT0121-100MG TCT0121-001G TCT0121-5G
Fisetin ≥96.0%
Fisetin

CAS Number: 528-48-3
MDL Number: MFCD00006829
Molecular Formula: C15H10O6
Molecular Weight: 286.24
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 330


Product Categories: Bioscience, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 3,7,3',4'-Tetrahydroxyflavone

Formula: C₁₅H₁₀O₆
Melting Pt: 330 °C
MDL Number: MFCD00006829
CAS Number: 528-48-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83eaf6c15eaed51950a1f383db5c76eb6df3815a&vid=83eaf6c15eaed51950a1f383db5c76eb6df3815a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR