Tetrahydrofurfuryl acetate ≥97.0%

Supplier: TCI America

Synonyms: Tetrahydrofurfuryl Acetate

T0105-25G T0105-500G
TCT0105-025GEA 113.76 CAD
TCT0105-025G TCT0105-500G
Tetrahydrofurfuryl acetate ≥97.0%
Tetrahydrofurfuryl acetate

CAS Number: 637-64-9
MDL Number: MFCD00022478
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 195
Flash Point (°C): 84
Specific Gravity (20/20): 1.06

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 195 °C
Density: 1.06 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00022478
CAS Number: 637-64-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7ad403e2892c2f4e210882a0d301b29cd3de0a4&vid=c7ad403e2892c2f4e210882a0d301b29cd3de0a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR