Terpinyl acetate ≥85.0%
Supplier: TCI America
T0023-25ML
T0023-500ML
TCT0023-025MLEA
27.33
CAD
TCT0023-025ML
TCT0023-500ML
Terpinyl acetate ≥85.0%
Terpinyl acetate
CAS Number: 80-26-2
MDL Number: MFCD00037155
Molecular Formula: C12H20O2
Molecular Weight: 196.29
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 94
Flash Point (°C): 115
Specific Gravity (20/20): 0.96
Formula:
C₁₂H₂₀O₂ Boiling Pt: 94 °C Density: 0.96 (20 °C) Flash Pt: 115 °C |
MDL Number:
MFCD00037155 CAS Number: 80-26-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...