Methyl-4-formylbenzoate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl terephthalaldehydate, Methyl 4-formylbenzoate
T0012-500G
T0012-25G
T0012-100G
TCT0012-500GEA
739.44
CAD
TCT0012-500G
TCT0012-025G
TCT0012-100G
Methyl-4-formylbenzoate ≥98.0%
Methyl 4-formylbenzoate
CAS Number: 1571-08-0
MDL Number: MFCD00006950
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 135
Melting point (°C): 60
Other Product Names / Synonyms: 4-Formylbenzoic Acid Methyl Ester, Methyl 4-Formylbenzoate, Terephthalaldehydic Acid Methyl Ester
Formula:
C₉H₈O₃ MW: 164.16 g/mol Boiling Pt: 135 °C Melting Pt: 60 °C |
MDL Number:
MFCD00006950 CAS Number: 1571-08-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...