Sodium 4-Hydroxybenzenesulfonate dihydrate ≥98.0%
Supplier: TCI America
Synonyms:
4-Phenolsulphonic acid sodium salt dihydrate, 4-Hydroxybenzenesulphonic acid sodium salt, p-Phenolsulfonic acid sodium salt, 4-Hydroxybenzenesulfonic acid sodium salt, Sodium 4-hydroxybenzenesulfonate dihydrate, Sodium 4-hydroxybenzenesulphonate dihydrate, Sodium p-Phenolsulphonate, p-Phenolsulphonic acid sodium salt, Sodium p-Phenolsulfonate, 4-Phenolsulfonic acid sodium salt dihydrate
CAS Number: 10580-19-5
MDL Number: MFCD00150724
Molecular Formula: C6H6O4S
Molecular Weight: 196.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Other Product Names / Synonyms: 4-Hydroxybenzenesulfonic Acid Sodium Salt, Sodium p-Phenolsulfonate, p-Phenolsulfonic Acid Sodium Salt
Formula:
C₆H₉NaO₆S MW: 232.19 g/mol |
MDL Number:
MFCD00150724 CAS Number: 10580-19-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...