Methyl-2-amino-3-hydroxypropanoate hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: Methyl DL-serinate hydrochloride, H-DL-Ser-OMe·HCl, 3-hydroxy-1-methoxy-1-oxopropan-2-aminium chloride, methyl 2-amino-3-hydroxypropanoate, DL-Serine Methyl Ester Hydrochloride, H-DL-Ser-OMe.HCl

S0576-25G S0576-5G
TCS0576-25GEA 48.98 CAD
TCS0576-25G TCS0576-5G
Methyl-2-amino-3-hydroxypropanoate hydrochloride ≥98.0%
Methyl-2-amino-3-hydroxypropanoate hydrochloride

CAS Number: 5619-04-5
MDL Number: MFCD00012593
Molecular Formula: C4H9NO3
Molecular Weight: 155.58
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Color: White
Melting point (°C): 136


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-DL-Ser-OMe.HCl

Formula: C₄H₁₀ClNO₃
MW: 155.578 g/mol
Melting Pt: 136 °C
MDL Number: MFCD00012593
CAS Number: 5619-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40462c3a28684b345d589f9cfee83c0f42017a1d&vid=40462c3a28684b345d589f9cfee83c0f42017a1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR