Sodium butyrate ≥98.0%

Supplier: TCI America

Synonyms: Butyric acid sodium salt, Sodium Butyrate

S0519-25G S0519-100G
TCS0519-25GEA 45.55 CAD
TCS0519-25G TCS0519-100G
Sodium butyrate ≥98.0%
Sodium butyrate

CAS Number: 156-54-7
MDL Number: MFCD00002816
Molecular Formula: C4H8O2
Molecular Weight: 110.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 253


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Lipids, Epigenetics Research Reagents
Other Product Names / Synonyms: Butyric Acid Sodium Salt

Formula: C₄H₇NaO₂
Melting Pt: 253 °C
MDL Number: MFCD00002816
CAS Number: 156-54-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97ba298b6e63d714d5742bfa5e381697df0641b1&vid=97ba298b6e63d714d5742bfa5e381697df0641b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR