Methyl-3-sulfolene-3-carboxylate ≥98.0%
Supplier: TCI America
Synonyms:
2,5-Dihydro-3-methoxycarbonylthiophene 1,1-Dioxide, 3-sulpholene-3-carboxylic acid Methyl Ester, Methyl 2,5-dihydrothiophene-3-carboxylate 1,1-dioxide, 3-Sulfolene-3-carboxylic acid Methyl Ester, 3-Methoxycarbonyl-3-sulfolene, 3-Methoxycarbonyl-3-sulpholene
CAS Number: 67488-50-0
MDL Number: MFCD00042754
Molecular Formula: C6H8O4S
Molecular Weight: 176.19
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 64
Other Product Names / Synonyms: 2,5-Dihydro-3-methoxycarbonylthiophene 1,1-Dioxide, 3-Methoxycarbonyl-3-sulfolene, 3-Sulfolene-3-carboxylic Acid Methyl Ester
Formula:
C₆H₈O₄S MW: 176.19 g/mol Melting Pt: 64 °C |
MDL Number:
MFCD00042754 CAS Number: 67488-50-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...