4-Sulfocalix[6]arene hydrate ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
Hexahydroxycalix[6]arenehexasulfonic acid, Hexahydroxycalix[6]arenehexasulphonic acid, Hexasulfo(hexahydroxy)calix[6]arene, Hexasulpho(hexahydroxy)calix[6]arene
CAS Number: 102088-39-1
MDL Number: MFCD00143376
Molecular Formula: C42H36O24S6
Molecular Weight: 1117.09
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 246
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: Hexahydroxycalix[6]arenehexasulfonic Acid, Hexasulfo(hexahydroxy)calix[6]arene
Formula:
C₄₂H₃₆O₂₄S₆ MW: 1117.09 g/mol Melting Pt: 246 °C |
MDL Number:
MFCD00143376 CAS Number: 102088-39-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...