Monomethyl suberate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

S0440-5G S0440-25G
TCS0440-005GEA 57.83 CAD
TCS0440-005G TCS0440-025G
Monomethyl suberate ≥98.0% (by GC, titration analysis)
Monomethyl suberate

CAS Number: 3946-32-5
MDL Number: MFCD00004427
Molecular Formula: C9H16O4
Molecular Weight: 188.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 135
Flash Point (°C): 110
Freezing point (°C): 21


Other Product Names / Synonyms: Methyl Hydrogen Suberate, Octanedioic Acid Monomethyl Ester, Suberic Acid Monomethyl Ester

Formula: C₉H₁₆O₄
Boiling Pt: 135 °C
Flash Pt: 110 °C
MDL Number: MFCD00004427
CAS Number: 3946-32-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dcdecf936468160d3e397da96fdc4994b9ee7bb&vid=1dcdecf936468160d3e397da96fdc4994b9ee7bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR