3-Nitrobenzyl alcohol ≥99.0%

Supplier: TCI America

Synonyms: (3-Nitrophenyl)methanol

S0378-1G S0378-10G
TCS0378-1GEA 34.76 CAD
TCS0378-1G TCS0378-10G
3-Nitrobenzyl alcohol ≥99.0%
3-Nitrobenzyl alcohol

[Matrix for FABMS and liquid SIMS]
CAS Number: 619-25-0
MDL Number: MFCD00007273
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Flash Point (°C): 110
Freezing point (°C): 30

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00007273
CAS Number: 619-25-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab3af97e9e63692fca90a8a4aa05f90bfdd9be93&vid=a8d4b34f6fca5018cf73e6ade894c6931836dd47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR