Salcomine [=N,N'-bis(salicylidene)ethylenediiminocobalt(II)] ≥95.0%
Supplier: TCI America
Synonyms:
N,N'-Disalicylalethylenediamine Cobalt(II), Salcomine, N,N'-Bis(salicylidene)ethylenediiminocobalt(II), N,N'-Ethylenebis(salicylideneiminato)cobalt(II)
CAS Number: 14167-18-1
MDL Number: MFCD00000009
Molecular Formula: C16H14CoN2O2
Molecular Weight: 325.23
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: Yellow Red
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Oxidation, Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: N,N'-Bis(salicylidene)ethylenediiminocobalt(II), N,N'-Disalicylalethylenediamine Cobalt(II), N,N'-Ethylenebis(salicylideneiminato)cobalt(II)
Formula:
C₁₆H₁₆CoN₂O₂ MW: 327.24 g/mol |
MDL Number:
MFCD00000009 CAS Number: 14167-18-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...