Salicylideneaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

S0267-25G S0267-5G
TCS0267-025GEA 137.46 CAD
TCS0267-025G TCS0267-5G
Salicylideneaniline ≥98.0% (by GC, titration analysis)
Salicylideneaniline

CAS Number: 779-84-0
MDL Number: MFCD00020066
Molecular Formula: C13H11NO
Molecular Weight: 197.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Yellow
Boiling point (°C): 151
Melting point (°C): 51


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: N-Salicylalaniline

Formula: C₁₃H₁₁NO
Boiling Pt: 151 °C
Melting Pt: 51 °C
MDL Number: MFCD00020066
CAS Number: 779-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc56904fd1f3886337e889c6891763a5b920e3fb&vid=cc56904fd1f3886337e889c6891763a5b920e3fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR