Sodium-p-styrenesulfonate hydrate ≥93.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: p-Styrenesulfonic acid sodium salt hydrate, p-Styrenesulphonic acid sodium salt hydrate, Sodium-p-styrenesulphonate hydrate, Sodium 4-vinylbenzenesulphonate hydrate, Sodium-p-styrenesulfonate hydrate, Styrene-4-sulfonic acid sodium salt hydrate, Sodium 4-vinylbenzenesulfonate hydrate, Styrene-4-sulphonic acid sodium salt hydrate

S0258-25G S0258-500G S0258-100G
TCS0258-025GEA 55.92 CAD
TCS0258-025G TCS0258-500G TCS0258-100G
Sodium-p-styrenesulfonate hydrate ≥93.0% (by titrimetric analysis)
Sodium-p-styrenesulfonate hydrate

CAS Number: 123333-94-8
MDL Number: MFCD00013379
Molecular Formula: C8H8O3S
Molecular Weight: 206.19
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Color: White


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: p-Styrenesulfonic Acid Sodium Salt

Formula: C₈H₇NaO₃S·xH₂O
MW: 206.19 g/mol (anhy)
MDL Number: MFCD00013379
CAS Number: 123333-94-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1a9ea78d6060cd196fcf69caaad505a51ddc070&vid=a1a9ea78d6060cd196fcf69caaad505a51ddc070&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR