DMS (Dimethyl suberimidate dihydrochloride) ≥96.0%

Supplier: TCI America

Synonyms: Suberimidic acid dimethyl ester dihydrochloride, DMS, Dimethyl ester octanediimidic acid dihydrochloride

S0246-5G S0246-25G
TCS0246-005GEA 136.67 CAD
TCS0246-005G TCS0246-025G
DMS (Dimethyl suberimidate dihydrochloride) ≥96.0%
DMS (Dimethyl suberimidate dihydrochloride)

[Cross-linking Agent for Protein Research]
CAS Number: 34490-86-3
MDL Number: MFCD00012574
Molecular Formula: C10H20N2O2
Molecular Weight: 273.20
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: White
Melting point (°C): 221
Storage Temperature: 0-10°C


Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: Suberimidic Acid Dimethyl Ester Dihydrochloride

Formula: C₁₀H₂₀N₂O₂
MW: 273.20 g/mol
Melting Pt: 221 °C
MDL Number: MFCD00012574
CAS Number: 34490-86-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab15fc9ccb6334f8f70b966d133466b9dd97b0c4&vid=ab15fc9ccb6334f8f70b966d133466b9dd97b0c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR