p-Sexiphenyl

Supplier: TCI America

S0220-100MG S0220-1G
TCS0220-100MGEA 186.76 CAD
TCS0220-100MG TCS0220-1G
p-Sexiphenyl
p-Sexiphenyl

CAS Number: 4499-83-6
MDL Number: MFCD00039559
Molecular Formula: C36H26
Molecular Weight: 458.60
Form: Powder
Color: White

Formula: C₃₆H₂₆
MDL Number: MFCD00039559
CAS Number: 4499-83-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=656562f11571394b526a98873589b9e384d4c336&vid=656562f11571394b526a98873589b9e384d4c336&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR