Monosodium 5-sulfoisophthalate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

S0148-25G S0148-500G S0148-100G
TCS0148-025GEA 30.02 CAD
TCS0148-025G TCS0148-500G TCS0148-100G
Monosodium 5-sulfoisophthalate ≥98.0% (by titrimetric analysis)
Monosodium 5-sulfoisophthalate

CAS Number: 6362-79-4
MDL Number: MFCD00007495
Molecular Formula: C8H6O7S
Molecular Weight: 268.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: 5-Sulfoisophthalic Acid Monosodium Salt, Sodium 5-Sulfoisophthalate

Formula: C₈H₆O₇S
MDL Number: MFCD00007495
CAS Number: 6362-79-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7eff7a7851043b7ba58d14d640b2dc8fb826cf9&vid=f7eff7a7851043b7ba58d14d640b2dc8fb826cf9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR