3-Sulfobenzoic acid monosodium salt ≥95.0%

Supplier: TCI America

Synonyms: Sodium 3-sulphobenzoate, Sodium 3-sulfobenzoate

S0126-500G S0126-25G
TCS0126-500GEA 249.64 CAD
TCS0126-500G TCS0126-025G
3-Sulfobenzoic acid monosodium salt ≥95.0%
3-Sulphobenzoic acid monosodium salt

CAS Number: 17625-03-5
MDL Number: MFCD00066378
Molecular Formula: C7H6O5S
Molecular Weight: 224.16
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White

Formula: C₇H₅NaO₅S
MW: 224.17 g/mol
MDL Number: MFCD00066378
CAS Number: 17625-03-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21c87a6eca439d0c2256e7513a5435fe83b4420a&vid=21c87a6eca439d0c2256e7513a5435fe83b4420a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR