Salicylohydroxamic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

S0008-5G S0008-25G
TCS0008-005GEA 37.92 CAD
TCS0008-005G TCS0008-025G
Salicylohydroxamic acid ≥98.0% (by HPLC, titration analysis)
Salicylohydroxamic acid

CAS Number: 89-73-6
MDL Number: MFCD00002110
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 173


Other Product Names / Synonyms: N,2-Dihydroxybenzamide, 2-Hydroxybenzohydroxamic Acid, N-Hydroxysalicylamide, Salicylohydroxamic Acid

Formula: C₇H₇NO₃
Melting Pt: 173 °C
MDL Number: MFCD00002110
CAS Number: 89-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b79d742b9dfe2cc59f0be470457247a528caa14f&vid=b79d742b9dfe2cc59f0be470457247a528caa14f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR