Ruthenium 5% on carbon wetted with ca. 40% water

Supplier: TCI America

R0076-5G R0076-25G
TCR0076-5GEA 90.06 CAD
TCR0076-5G TCR0076-25G
Ruthenium 5% on carbon wetted with ca. 40% water
Ruthenium

CAS Number: 7440-18-8
Molecular Formula: Ru
Molecular Weight: 101.07
Form: Crystals
Color: Black


Product Categories: Synthetic Organic Chemistry, Functional Group Transformation and Introducing Reaction, Metal Catalysts for Organic Synthesis

Formula: Ru
MDL Number: MFCD00011207
CAS Number: 7440-18-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6688d02a65d17ec0b4314c3ac3042a5364d816d3&vid=f540613b2964f84192f95d1004520cf5845220c4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR