β-D-Ribofuranose-1,2,3,5-tetraacetate ≥98.0%
Supplier: TCI America
Synonyms:
Tetra-O-acetyl-β-D-ribofuranose, (2S,3R,4R,5R)-5-(Acetoxymethyl)tetrahydrofuran-2,3,4-triyl triacetate
CAS Number: 13035-61-5
MDL Number: MFCD00005358
Molecular Formula: C13H18O9
Molecular Weight: 318.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 83
Specific rotation [a]20/D: -14.5 deg (C=5, MeOH)
Product Categories: Synthetic Organic Chemistry, Glycosylation, Nucleic Acid Synthesis, Bioscience, Ribose, O-Substituted Sugars
Other Product Names / Synonyms: beta-D-Ribofuranose 1,2,3,5-Tetraacetate
Formula:
C13H18O9 MW: 318.28 g/mol Melting Pt: 83 °C |
MDL Number:
MFCD00005358 CAS Number: 13035-61-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...