3-Quinolineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Q0080-1G
Q0080-5G
TCQ0080-1GEA
85.32
CAD
TCQ0080-1G
TCQ0080-5G
3-Quinolineboronic acid (contains varying amounts of Anhydride)
3-Quinolineboronic acid
CAS Number: 191162-39-7
MDL Number: MFCD02183527
Molecular Formula: C9H8BNO2
Molecular Weight: 172.98
Form: Crystal
Color: White
Formula:
C₉H₈BNO₂ |
MDL Number:
MFCD02183527 CAS Number: 191162-39-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...