6-Quinolinecarboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Q0077-25G Q0077-5G
TCQ0077-25GEA 553 CAD
TCQ0077-25G TCQ0077-5G
6-Quinolinecarboxylic acid ≥98.0% (by HPLC, titration analysis)
6-Quinolinecarboxylic acid

CAS Number: 10349-57-2
MDL Number: MFCD00047613
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 291

Formula: C₁₀H₇NO₂
Melting Pt: 291 °C
MDL Number: MFCD00047613
CAS Number: 10349-57-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b34f2fb29c051b9ad146d062462302a54ecff15&vid=40f3664ea2c6bc68896e6889e7690e3b04ad2dee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR