3-Quinolinecarbaldehyde ≥96.0% (by GC, titration analysis)

Supplier: TCI America

Q0075-1G Q0075-5G
TCQ0075-1GEA 161.16 CAD
TCQ0075-1G TCQ0075-5G
3-Quinolinecarbaldehyde ≥96.0% (by GC, titration analysis)
3-Quinolinecarbaldehyde

CAS Number: 13669-42-6
MDL Number: MFCD00006768
Molecular Formula: C10H7NO
Molecular Weight: 157.17
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 128
Melting point (°C): 67
Lambda max.: 282 nm (MeOH)


Other Product Names / Synonyms: 3-Formylquinoline

Formula: C₁₀H₇NO
Boiling Pt: 128 °C
Melting Pt: 67 °C
MDL Number: MFCD00006768
CAS Number: 13669-42-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebf08128e2d3164d850a1d415cf8bbedf5fe591e&vid=7724329c1387547c596cba69fdc80b2ddb03be39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR