4-Quinolinecarboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: Quinoline-4-carboxylic acid, NSC 13138

Q0065-25G Q0065-1G Q0065-5G
TCQ0065-25GEA 1225.07 CAD
TCQ0065-25G TCQ0065-1G TCQ0065-5G
4-Quinolinecarboxylic acid ≥98.0%
4-Quinolinecarboxylic acid

CAS Number: 486-74-8
MDL Number: MFCD00006782
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
MDL Number: MFCD00006782
CAS Number: 486-74-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec95fe087addb02d6fa65d669eed636bdd478e66&vid=d20ce6ab38d6d7658b49a15829b48b84c632ee95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR