4-Propylcyclohexanone ≥98.0%

Supplier: TCI America

P1849-25G P1849-5G
TCP1849-25GEA 88.43 CAD
TCP1849-25G TCP1849-5G
4-Propylcyclohexanone ≥98.0%
4-Propylcyclohexanone

CAS Number: 40649-36-3
MDL Number: MFCD00058690
Molecular Formula: C9H16O
Molecular Weight: 140.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 98
Flash Point (°C): 86
Specific Gravity (20/20): 0.91

Formula: C₉H₁₆O
Boiling Pt: 98 °C
Density: 0.91 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00058690
CAS Number: 40649-36-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93c0467297c74cc829dbc72bd69aa1868b9c6dce&vid=93c0467297c74cc829dbc72bd69aa1868b9c6dce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR