Potassium(4-methoxyphenyl)trifluoroborate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(4-Methoxyphenyl)trifluoroboric acid potassium salt, Potassium (4-methoxyphenyl)trifluoroborate, Potassium trifluoro(4-methoxyphenyl)borate
P1820-5G
P1820-1G
TCP1820-5GEA
228.15
CAD
TCP1820-5G
TCP1820-1G
Potassium(4-methoxyphenyl)trifluoroborate ≥98.0% (by HPLC, titration analysis)
Potassium (4-methoxyphenyl)trifluoroborate
CAS Number: 192863-36-8
MDL Number: MFCD04112717
Molecular Formula: C7H7BF3KO
Molecular Weight: 214.04
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Formula:
C₇H₇BF₃KO MW: 214.04 g/mol |
MDL Number:
MFCD04112717 CAS Number: 192863-36-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...