(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer ≥96.0% (by titrimetric analysis)

Supplier: TCI America

P1788-200MG P1788-1G
TCP1788-200MGEA 257.54 CAD
TCP1788-200MG TCP1788-1G
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer ≥96.0% (by titrimetric analysis)
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer

CAS Number: 12354-85-7
MDL Number: MFCD00061552
Molecular Formula: C20H30Cl4Rh2
Molecular Weight: 618.07
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: Yellow


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, C-H Bond Activation Reaction
Other Product Names / Synonyms: Dichloro(pentamethylcyclopentadienyl)rhodium(III) Dimer

Formula: C₂₀H₃₀Cl₄Rh₂
MDL Number: MFCD00061552
CAS Number: 12354-85-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89d4a594d6d9ce165a442e7fafc4aeb7e272e939&vid=89d4a594d6d9ce165a442e7fafc4aeb7e272e939&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR