Palladium 10% on carbon wetted with ca. 55% water

Supplier: TCI America

P1785-25G P1785-5G
TCP1785-25GEA 967.59 CAD
TCP1785-25G TCP1785-5G
Palladium 10% on carbon wetted with ca. 55% water
Palladium

CAS Number: 7440-05-3
Molecular Formula: Pd
Molecular Weight: 106.42
Form: Crystals
Color: Black


Useful catalyst for coupling reaction, etc.
Product Categories: Synthetic Organic Chemistry, Functional Group Transformation and Introducing Reaction, Metal Catalysts for Organic Synthesis, Suzuki-Miyaura Cross-coupling Reaction

Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8941a2fcda34204b15666f06dacfcacaf4ad5c48&vid=5faef58c06ebe23d49ab4a129120985fc13c6aa6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR