N-Phenylglycine ethyl ester ≥98.0%

Supplier: TCI America

P1781-25G P1781-5G
TCP1781-25GEA 388.36 CAD
TCP1781-25G TCP1781-5G
N-Phenylglycine ethyl ester ≥98.0%
N-Phenylglycine ethyl ester

CAS Number: 2216-92-4
MDL Number: MFCD00038315
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 140
Melting point (°C): 56


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: Ethyl Anilinoacetate, Anilinoacetic Acid Ethyl Ester

Formula: C₁₀H₁₃NO₂
Boiling Pt: 140 °C
Melting Pt: 56 °C
MDL Number: MFCD00038315
CAS Number: 2216-92-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93c7e9b33c57d69dfc1f02304071f835bd4b41d5&vid=93c7e9b33c57d69dfc1f02304071f835bd4b41d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR