Polyethylene glycol monododecyl ether, M.W. n=∼25 for biochemical research

Supplier: TCI America

P1777-25G P1777-5G
TCP1777-25GEA 47.4 CAD
TCP1777-25G TCP1777-5G
Polyethylene glycol monododecyl ether, M.W. n=∼25 for biochemical research
Polyethylene glycol monododecyl ether

[for Biochemical Research]
CAS Number: 9002-92-0
MDL Number: MFCD00080891
Form: Crystal
Color: White
Flash Point (°C): 170


Other Product Names / Synonyms: Polyethylene Glycol Monolauryl Ether

Formula: HO(CH₂CH₂O)nC₁₂H₂₅
Flash Pt: 170 °C
MDL Number: MFCD00080891
CAS Number: 9002-92-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b63b93ffeeac370156291a58fe0e8ed919e66839&vid=f8961edecc6d3e4bcf66e1e8c5cb7ff5665ed258&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR