N,N,2,4,6-Pentamethylaniline ≥97.0%

Supplier: TCI America

Synonyms: 2,4,6,N,N-Pentamethylaniline

P1767-25G P1767-5G
TCP1767-25GEA 240.16 CAD
TCP1767-25G TCP1767-5G
N,N,2,4,6-Pentamethylaniline ≥97.0%
N,N,2,4,6-Pentamethylaniline

CAS Number: 13021-15-3
MDL Number: MFCD00010596
Molecular Formula: C11H17N
Molecular Weight: 163.26
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 215
Flash Point (°C): 79
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: N,N-Dimethylmesidine, 2-(Dimethylamino)mesitylene

Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Boiling Pt: 215 °C
Density: 0.91 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00010596
CAS Number: 13021-15-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a1761710aaa66325391208ddc40f79cb3134eaf&vid=5a1761710aaa66325391208ddc40f79cb3134eaf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR