1H,1H-Pentadecafluoro-n-octyl acrylate ≥95.0% stabilized
Supplier: TCI America
Synonyms:
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluorooctyl acrylate, Acrylic Acid 1H,1H-Pentadecafluoro-n-octyl Ester
(stabilized with MEHQ)
CAS Number: 307-98-2
MDL Number: MFCD00039248
Molecular Formula: C11H5F15O2
Molecular Weight: 454.14
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 65
Flash Point (°C): 91
Specific Gravity (20/20): 1.62
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid 1H,1H-Pentadecafluoro-n-octyl Ester
Formula:
C₁₁H₅F₁₅O₂ MW: 454.14 g/mol Boiling Pt: 65 °C Density: 1.62 (20 °C) Flash Pt: 91 °C |
MDL Number:
MFCD00039248 CAS Number: 307-98-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...