D-Proline methyl ester hydrochloride ≥96.0%

Supplier: TCI America

Synonyms: H-D-Pro-OMe.HCl

P1730-1G P1730-5G
TCP1730-1GEA 47.4 CAD
TCP1730-1G TCP1730-5G
D-Proline methyl ester hydrochloride ≥96.0%
D-Proline methyl ester hydrochloride

CAS Number: 65365-28-8
MDL Number: MFCD00083685
Molecular Formula: C6H11NO2
Molecular Weight: 165.62
Purity/Analysis Method: >96.0% (N,T)
Form: Crystal
Color: White
Melting point (°C): 73
Specific rotation [a]20/D: 32 deg (C=1, H2O)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Pro-OMe.HCl

Formula: C₆H₁₁NO₂
MW: 165.62 g/mol
Melting Pt: 73 °C
MDL Number: MFCD00083685
CAS Number: 65365-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4ad24e40544b0932aff8fd7a581bb3ace401cf6&vid=a4ad24e40544b0932aff8fd7a581bb3ace401cf6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR