L-Proline benzyl ester hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
H-Pro-OBzl.HCl
P1727-25G
P1727-5G
TCP1727-25GEA
105.86
CAD
TCP1727-25G
TCP1727-5G
L-Proline benzyl ester hydrochloride ≥98.0%
L-Proline benzyl ester hydrochloride
CAS Number: 16652-71-4
MDL Number: MFCD00039002
Molecular Formula: C12H15NO2
Molecular Weight: 241.72
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 144
Specific rotation [a]20/D: -43 deg (C=1, MeOH)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Pro-OBzl.HCl
Formula:
C₁₂H₁₅NO₂ MW: 241.72 g/mol Melting Pt: 144 °C |
MDL Number:
MFCD00039002 CAS Number: 16652-71-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...