D-Phenylalanine methyl ester hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: (R)-Methyl 2-amino-3-phenylpropanoate hydrochloride, H-D-Phe-OMe.HCl, H-D-Phe-OMe·HCl

P1725-25G P1725-5G
TCP1725-25GEA 118.5 CAD
TCP1725-25G TCP1725-5G
D-Phenylalanine methyl ester hydrochloride ≥98.0%
D-Phenylalanine methyl ester hydrochloride

CAS Number: 13033-84-6
MDL Number: MFCD00066112
Molecular Formula: C10H13NO2
Molecular Weight: 215.68
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 160
Specific rotation [a]20/D: -37 deg (C=2, EtOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Phe-OMe.HCl

Formula: C₁₀H₁₃NO₂
MW: 215.68 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00066112
CAS Number: 13033-84-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a39fc5637ce04a45905efaedf7a504a27e1ccdf3&vid=a39fc5637ce04a45905efaedf7a504a27e1ccdf3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR