L-Phenylalanine benzyl ester hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
P1718-25G
P1718-5G
TCP1718-25GEA
199.08
CAD
TCP1718-25G
TCP1718-5G
L-Phenylalanine benzyl ester hydrochloride ≥98.0% (by HPLC, total nitrogen)
L-Phenylalanine benzyl ester hydrochloride
CAS Number: 2462-32-0
MDL Number: MFCD00043249
Molecular Formula: C16H17NO2
Molecular Weight: 291.78
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 203
Specific rotation [a]20/D: -12 deg (C=1, AcOH(80%))
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Phe-OBzl.HCl
Formula:
C₁₆H₁₇NO₂ Melting Pt: 203 °C |
MDL Number:
MFCD00043249 CAS Number: 2462-32-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...