Palladium 5% on calcium carbonate for use as a catalyst lead poisoned
Supplier: TCI America
P1703-25G
P1703-5G
TCP1703-25GEA
1219.76
CAD
TCP1703-25G
TCP1703-5G
Palladium 5% on calcium carbonate for use as a catalyst lead poisoned
Palladium
CAS Number: 7440-05-3
Molecular Formula: Pd
Molecular Weight: 106.42
Form: Crystals
Color: Grey
Product Categories: Synthetic Organic Chemistry, Functional Group Transformation and Introducing Reaction, Metal Catalysts for Organic Synthesis
Formula:
Pd MW: 106.420 g/mol |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...