Palladium 5% on barium sulphate

Supplier: TCI America

P1702-25G P1702-5G
TCP1702-25GEA 1060.18 CAD
TCP1702-25G TCP1702-5G
Palladium 5% on barium sulphate
Palladium

CAS Number: 7440-05-3
Molecular Formula: Pd
Molecular Weight: 106.42
Form: Crystals
Color: Grey


Product Categories: Synthetic Organic Chemistry, Functional Group Transformation and Introducing Reaction, Metal Catalysts for Organic Synthesis

Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb418bbfdc61fdf387e06045aeff87e4292f882c&vid=4b518ba61cf3ffdd3e2e85d3d490afdccba81f12&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR