2-Propoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P1694-5G P1694-25G
TCP1694-5GEA 41.08 CAD
TCP1694-5G TCP1694-25G
2-Propoxybenzoic acid ≥98.0% (by GC, titration analysis)
2-Propoxybenzoic acid

CAS Number: 2100-31-4
MDL Number: MFCD01075687
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 207
Melting point (°C): 39
Storage Temperature: 0-10°C

Formula: C₁₀H₁₂O₃
Boiling Pt: 207 °C
Melting Pt: 39 °C
MDL Number: MFCD01075687
CAS Number: 2100-31-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ff2611f575a1ec527ff489c9a7dc91bcda3e76f&vid=4ff2611f575a1ec527ff489c9a7dc91bcda3e76f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR